Anti-PLVAP Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: MECA-32]
Catalog # USBIP4256-25V
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Plasmalemma Vesicle Associated Protein
- Antigen symbol:PLVAP
- Clonality:Monoclonal
- Clone:MECA-32
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2a
- Reactivity:Mouse
- Form:liquid
- Storage buffer:PBS, pH 7,4, 1% BSA, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Flow Cytometry: 1:5-1:10;
Type: Primary
Antigen: PLVAP (plasmalemma vesicle associated protein)
Clonality: monoclonal
Clone: MECA-32
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse