Anti-Phosphoinositide 3 kinase, p110 delta Rabbit Polyclonal Antibody
Catalog # USBIP4073-26
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Phosphoinositide 3 Kinase, p110 delta
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:1026 to 1044
- Storage buffer:PBSbuffer, 1mg/ml BSA, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:3000; Immunochemistry: 1:400; ELISA: 1:1000
Type: Primary
Antigen: Phosphoinositide 3 kinase, p110 delta
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human