Anti-PED/PEA-15 Rabbit Polyclonal Antibody
Catalog # USBIP3139-85B
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:PED
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Antigen synonyms:PEA-15
- Modification:phospho-Ser116
- Storage buffer:PBS(without Mg2+ and Ca2+), pH 7,2, 1mg/mlBSA (IgG, protease free), 0,05% sodium azide, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:1000
Type: Primary
Antigen: PED/PEA-15
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat