Anti-PARP 10 Rat Monoclonal Antibody [clone: 6D643]
Catalog # USBIP3113-45
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:PARP10
- Clonality:Monoclonal
- Clone:6D643
- Conjugation:Unconjugated
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Format:Supernatant
- Form:liquid
- Amino acid number:1 to 907
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Recommended dilution: Immunochemistry: 1:10; Immunoprecipitation: 1:10; Western Blot: 1:50
Type: Primary
Antigen: PARP 10
Clonality: monoclonal
Clone: 6D643
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human