Anti-Papilloma Virus, Human, Type 16 L1, Late Mouse Monoclonal Antibody [clone: 5i81]
Catalog # USBIP3105-31A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Papilloma Virus, Human, Type 16 L1, Late
- Clonality:Monoclonal
- Clone:5i81
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b
- Form:liquid
- Storage buffer:0,1M Tris-HCl, pH 7,5.
- Storage temperature:Cool
- Concentration:~0,2 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Papilloma Virus, Human, Type 16 L1, Late
Clonality: Monoclonal
Clone: 5i81
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: