Anti-Papilloma Virus Type 18 Mouse Monoclonal Antibody [clone: 9B106]
Catalog # USBIP3105-25A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Papilloma Virus Type 18
- Clonality:Monoclonal
- Clone:9B106
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~4 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Primary
Antigen: Papilloma Virus Type 18
Clonality: Monoclonal
Clone: 9B106
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human