Anti-Papilloma Virus, Human, Type 18, Early Mouse Monoclonal Antibody [clone: 10F271]
Catalog # USBIP3105-24B
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Papilloma Virus, Human, Type 18, Early
- Clonality:Monoclonal
- Clone:10F271
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:200 µg
- Pk:200 µG
Specifications
About this item
Type: Primary
Antigen: Papilloma Virus, Human, Type 18, Early
Clonality: Monoclonal
Clone: 10F271
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: