Anti-Papilloma Virus, Human, Type 18, E6 Mouse Monoclonal Antibody [clone: 9L397]
Catalog # USBIP3105-23B
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Papilloma Virus, Human, Type 18, E6
- Clonality:Monoclonal
- Clone:9L397
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µg
- Pk:200 µG
Specifications
About this item
Type: Primary
Antigen: Papilloma Virus, Human, Type 18, E6
Clonality: Monoclonal
Clone: 9L397
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: