Anti-Papilloma Virus, Human, Type 16, E7 Mouse Monoclonal Antibody [clone: 10F268]
USBIP3105-16H
- Antibody type:Primary
- Antigen name:Papilloma Virus, Human, Type 16, E7
- Clonality:Monoclonal
- Clone:10F268
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:200 µg
- Pk:200 µG
Type: Primary
Antigen: Papilloma Virus, Human, Type 16, E7
Clonality: Monoclonal
Clone: 10F268
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: