Anti-Papilloma Virus, Human, Type 16, E7 Mouse Monoclonal Antibody [clone: 9B105]
USBIP3105-16F
- Antibody type:Primary
- Antigen name:Papilloma Virus, Human, Type 16, E7
- Clonality:Monoclonal
- Clone:9B105
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~2 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:1 mg
- Pk:1 mg
Type: Primary
Antigen: Papilloma Virus, Human, Type 16, E7
Clonality: Monoclonal
Clone: 9B105
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human