Anti-Papilloma Virus, Human, Type 16, Early Mouse Monoclonal Antibody [clone: 5E733]
Catalog # USBIP3105-07D
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Papilloma Virus, Human, Type 16, Early
- Clonality:Monoclonal
- Clone:5E733
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,1% sodium azide, before the addition of glycerol to 40%.
- Storage temperature:Cool
- Concentration:~4 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:500 µg
- Pk:500 µG
Specifications
About this item
Type: Primary
Antigen: Papilloma Virus, Human, Type 16, Early
Clonality: Monoclonal
Clone: 5E733
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: