Anti-PACAP-38 Rabbit Polyclonal Antibody
Catalog # USBIP1775-03C
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:PACAP38
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Sheep,Human,Rat
- Form:lyophilized powder
- Antigen synonyms:PACAP-38
- Amino acid number:28 to 38
- Storage buffer:PBS
- Sequence:GKRYKQRVKNK-NH2
- Storage temperature:Cool
- Concentration:~1 mg/ml (after reconstitution)
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: 1:3200
Type: Primary
Antigen: PACAP-38
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat, Sheep