Anti-TP53 Rabbit Monoclonal Antibody [clone: 9H263]
Catalog # USBIP1001-32G6
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Tumor protein p53
- Antigen symbol:P53
- Clonality:Monoclonal
- Clone:9H263
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Format:Supernatant
- Form:liquid
- Storage buffer:50mM Tris-glycine, pH 7,4, 0,15M sodium chloride, 0,05% BSA, 0,01% sodium azide, 40% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Flow Cytometry: 1:50; Western Blot: 1:250-1:500; Immunoprecipitation: 1:500; Immunochemistry: 1:100
Type: Primary
Antigen: TP53 (tumor protein p53)
Clonality: monoclonal
Clone: 9H263
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human