Anti-LAT2 Mouse Monoclonal Antibody [clone: NAP-03]
Catalog # USBIN6000-26C4
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:linker for activation of T cells family, member 2
- Antigen symbol:LAT2
- Clonality:Monoclonal
- Clone:NAP-03
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:196 to 212
- Storage buffer:PBS, 15mM sodium azide, pH 7,4.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Western Blot: 1-2?g/ml; Immunochemistry: 5 ?g/ml; Immunoprecipitation: 1-2?g/ml
Type: Primary
Antigen: LAT2 (linker for activation of T cells family, member 2)
Clonality: monoclonal
Clone: NAP-03
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human