Anti-NOTCH3 Rabbit Polyclonal Antibody
Catalog # USBIN5375-26
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Notch 3
- Antigen symbol:NOTCH3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Form:liquid
- Amino acid number:2306 to 2321
- Storage buffer:PBS, 0,1% sodium azide, before the addition of glycerol to 40%.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: ELISA: 1:1000
Type: Primary
Antigen: NOTCH3 (notch 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human