Anti-NOD2 Mouse Monoclonal Antibody [clone: NOD-15]
USBIN3006-02A
- Antibody type:Primary
- Antigen name:Nucleotide binding oligomerization domain containing 2
- Antigen symbol:NOD2
- Clonality:Monoclonal
- Clone:NOD-15
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Western Blot: 5 ?g/5ml
Type: Primary
Antigen: NOD2 (nucleotide-binding oligomerization domain containing 2)
Clonality: monoclonal
Clone: NOD-15
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human