Anti-RELA Rabbit Polyclonal Antibody
Catalog # USBIN2302-01L
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Transcription factor p65
- Antigen symbol:RELA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS(without Mg2+ and Ca2+), pH 7,4, 150mMsodium chloride, 0,02% sodium azide and 50% glycerol.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 1:500-1:1000; Immunochemistry: 1:50-1:100
Type: Primary
Antigen: RELA (v-rel avian reticuloendotheliosis viral oncogene homolog A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human