Order Entry
Export
ContactUsLinkComponent
 
Anti-Neuropeptide B-29 Rabbit Polyclonal Antibody (Biotin)
  USBIN2175-91D1
Anti-Neuropeptide B-29 Rabbit Polyclonal Antibody (Biotin)
  USBIN2175-91D1

 

  • Antibody type:
    Primary
  • Antigen name:
    Neuropeptide B-29
  • Clonality:
    Polyclonal
  • Conjugation:
    Biotin
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Form:
    liquid
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Type: Primary
Antigen: Neuropeptide B-29
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse