Anti-Neuropeptide B-29 Rabbit Polyclonal Antibody (Carboxyfluorescein)
USBIN2175-91C2
- Antibody type:Primary
- Antigen name:Neuropeptide B-29
- Clonality:Polyclonal
- Conjugation:FAM (Carboxyfluorescein)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat
- Form:liquid
- Storage temperature:Cool
- Shipping temperature:Ice
- Tested applications:Fluoresence ELISA
- Purification:purified
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: Neuropeptide B-29
Clonality: polyclonal
Clone:
Conjugation: Carboxyfluorescein
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat