Anti-NHERF1 Mouse Monoclonal Antibody [clone: EBP-10]
Catalog # USBIN2025-02C
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:NHERF1
- Clonality:Monoclonal
- Clone:EBP-10
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,4, 15mM sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: NHERF1
Clonality: monoclonal
Clone: EBP-10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human