Anti-Nectin 2 Rat Monoclonal Antibody [clone: 502-57]
Catalog # USBIN0571-50C
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Nectin 2
- Antigen symbol:NECTIN2
- Clonality:Monoclonal
- Clone:502-57
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rat
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 0,1% BSA, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Flow Cytometry: 1:10; Western Blot: 1:10; Immunochemistry: 1:10
Type: Primary
Antigen: Nectin 2
Clonality: monoclonal
Clone: 502-57
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse