Order Entry
Export
ContactUsLinkComponent
 
Anti-MYH1 Mouse Monoclonal Antibody [clone: MY-32]
  USBIM9850-41
undefined
Anti-MYH1 Mouse Monoclonal Antibody [clone: MY-32]
  USBIM9850-41

 

  • Antibody type:
    Primary
  • Antigen name:
    myosin, heavy chain 1, skeletal muscle, adult
  • Antigen symbol:
    MYH1
  • Clonality:
    Monoclonal
  • Clone:
    MY-32
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,4.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Recommended dilution: Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: MYH1 (myosin, heavy chain 1, skeletal muscle, adult)
Clonality: monoclonal
Clone: MY-32
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human