Order Entry
Export
ContactUsLinkComponent
 
Anti-Mucin 12 Rabbit Polyclonal Antibody
  USBIM4701-82-100
Anti-Mucin 12 Rabbit Polyclonal Antibody
  USBIM4701-82-100

 

  • Antibody type:
    Primary
  • Antigen name:
    Mucin 12
  • Antigen symbol:
    MUC12
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Amino acid number:
    541 to 555
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: Mucin 12
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human