Order Entry
Export
ContactUsLinkComponent
Anti-ABCC5 Rat Monoclonal Antibody [clone: M5II-54]
Anti-ABCC5 Rat Monoclonal Antibody [clone: M5II-54]
Catalog # USBIM4688-27C
Supplier:  US Biological
Anti-ABCC5 Rat Monoclonal Antibody [clone: M5II-54]
Catalog # USBIM4688-27C
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ATP Binding Cassette, sub-family C, member 5
  • Antigen symbol:
    ABCC5
  • Clonality:
    Monoclonal
  • Clone:
    M5II-54
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Format:
    Supernatant
  • Form:
    liquid
  • Amino acid number:
    722 to 910
  • Storage buffer:
    0,7%BSA, 0,1% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~250 ug immunoglobulin/ml (by ELISA),
  • Shipping temperature:
    Ice
  • Size:
    1 ml
  • Pk:
    1 mL

Specifications

About this item

Recommended dilution: Immunochemistry: 1:20-50;; Western Blot: 1:20-50

Type: Primary
Antigen: ABCC5 (ATP-binding cassette, sub-family C (CFTR/MRP), member 5)
Clonality: monoclonal
Clone: M5II-54
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human