Anti-c-Met Rabbit Polyclonal Antibody
USBIM3007-16
- Antibody type:Primary
- Antigen symbol:c-Met
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:liquid
- Modification:phospho-Tyr1230, phospho-Tyr1234, phospho-Tyr1235
- Storage buffer:Dulbecco’s PBS (without Mg2+ and Ca2+), 1mg/ml BSA (IgG, protease free), pH 7,3, 0,05% sodium azide, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:10 Pieces
- Pk:10
Recommended dilution: Western Blot: 1:1000
Type: Primary
Antigen: c-Met
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse