Anti-MEK1, MEK2 Rabbit Polyclonal Antibody
USBIM2865-51
- Antibody type:Primary
- Antigen symbol:MEK1/2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:liquid
- Modification:phospho-Ser218, phospho-Ser222
- Storage buffer:10mM HEPES, pH 7,5, 0,01% BSA, 0,09% sodium azide, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: MEK1, MEK2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse