Anti-MAP2K1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Catalog # USBIM2865-12D-AP
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Mitogen-activated protein kinase kinase 1
- Antigen symbol:MAP2K1
- Clonality:Polyclonal
- Conjugation:AP (Alkaline Phosphatase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Modification:phospho-Ser217, phospho-Ser221
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Do not freeze!
Type: Primary
Antigen: MAP2K1 (mitogen-activated protein kinase kinase 1)
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human