Anti-MAP2K1 Rabbit Polyclonal Antibody
USBIM2865-11N
- Antibody type:Primary
- Antigen name:Mitogen-activated protein kinase kinase 1
- Antigen symbol:MAP2K1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Hamster,Rabbit,Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Modification:phospho-Thr292
- Storage buffer:PBS(without Mg2+ and Ca2+), pH 7,2, 1mg/mlBSA (IgG, protease free), 0,05% sodium azide, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µl
- Pk:100 µl
Recommended dilution: Western Blot: 1:1000
Type: Primary
Antigen: MAP2K1 (mitogen-activated protein kinase kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Hamster, Human, Mouse, Rabbit, Rat