Anti-MEK 1 Rabbit Polyclonal Antibody
Catalog # USBIM2865-11A1
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:MEK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Modification:phospho-Thr386
- Storage buffer:10mM HEPES, pH 7,5, 0,1% BSA, 0,09% BSA and50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:1000
Type: Primary
Antigen: MEK 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human