Order Entry
Export
ContactUsLinkComponent
 
Anti-MAP2K1 Rabbit Polyclonal Antibody
  USBIM2865-09D
Anti-MAP2K1 Rabbit Polyclonal Antibody
  USBIM2865-09D

 

  • Antibody type:
    Primary
  • Antigen name:
    Mitogen-activated protein kinase kinase 1
  • Antigen symbol:
    MAP2K1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS(without Mg2+ and Ca2+), pH 7,4, 150mMsodium chloride, 0,02% sodium azide, 50% glycerol.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

 

 

Recommended dilution: Western Blot: 1:500-1:1000; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: MAP2K1 (mitogen-activated protein kinase kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat