Anti-MEK1, MEK2 Rabbit Polyclonal Antibody
USBIM2865-02G
- Antibody type:Primary
- Antigen symbol:MEK1/2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Modification:phospho-Ser217, phospho-Ser221
- Storage buffer:PBS, 1% BSA, 0,02% sodium azide, 50% glycerol.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Recommended dilution: Western Blot: 1:200-1:1000; Immunochemistry: 1:100-1:200
Type: Primary
Antigen: MEK1, MEK2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat