Order Entry
Export
ContactUsLinkComponent
Anti-MEK1, MEK2 Mouse Monoclonal Antibody [clone: 6D577]
Anti-MEK1, MEK2 Mouse Monoclonal Antibody [clone: 6D577]
Catalog # USBIM2865-02D-100
Supplier:  US Biological
Anti-MEK1, MEK2 Mouse Monoclonal Antibody [clone: 6D577]
Catalog # USBIM2865-02D-100
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    MEK1/2
  • Clonality:
    Monoclonal
  • Clone:
    6D577
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human,
    Dog,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recommended dilution: Western Blot: 1:20

Type: Primary
Antigen: MEK1, MEK2
Clonality: monoclonal
Clone: 6D577
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Mouse, Rat