Order Entry
Export
ContactUsLinkComponent
 
Anti-MEK1, MEK2 Rabbit Polyclonal Antibody
  USBIM2865-02A1
Anti-MEK1, MEK2 Rabbit Polyclonal Antibody
  USBIM2865-02A1

 

  • Antibody type:
    Primary
  • Antigen symbol:
    MEK1/2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Drosophila,
    Chicken,
    Human,
    Bovine,
    Dog,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Amino acid number:
    200 to 250
  • Storage buffer:
    PBS, 0,05% BSA, 0,05% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,5 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Recommended dilution: Western Blot: 5 ?g/ml

Type: Primary
Antigen: MEK1, MEK2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Chicken, Dog, Drosophila, Human, Mouse, Rat