Anti-MDR Antibody Kit, BioAssay™ Mouse Monoclonal Antibody [clone: JSB-1, LRP-56, MRPm6, MRPr1]
Catalog # USBIM2825-02
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:MDR
- Clonality:Monoclonal
- Clone:JSB-1, LRP-56, MRPm6, MRPr1
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b,IgG2a,IgG1
- Reactivity:Human
- Western blot:Yes
- Format:Supernatant
- Form:liquid
- Storage buffer:PBS, 0,1% BSA, 0,1% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 Kit
- Pk:1 KIT
Specifications
About this item
Type: Primary
Antigen: MDR Antibody Kit, BioAssay™
Clonality: monoclonal
Clone: JSB-1, LRP-56, MRPm6, MRPr1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1+IgG2b+IgG1+IgG2a
Reactivity: Human