Order Entry
Export
ContactUsLinkComponent
Anti-LY86 Rat Monoclonal Antibody [clone: 10F70]
Anti-LY86 Rat Monoclonal Antibody [clone: 10F70]
Catalog # USBIM2767-01C
Supplier:  US Biological
Anti-LY86 Rat Monoclonal Antibody [clone: 10F70]
Catalog # USBIM2767-01C
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    lymphocyte antigen 86
  • Antigen symbol:
    LY86
  • Clonality:
    Monoclonal
  • Clone:
    10F70
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG2b
  • Reactivity:
    Mouse
  • Form:
    liquid
  • Storage buffer:
    PBS, 0,09% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,5 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified
  • Size:
    250 µg
  • Pk:
    250 µG

Specifications

About this item

Recommended dilution: Flow Cytometry: 1:10-1:50;

Type: Primary
Antigen: LY86 (lymphocyte antigen 86)
Clonality: monoclonal
Clone: 10F70
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse