Order Entry
Export
ContactUsLinkComponent
Anti-Matrix Metalloproteinase 15 Rabbit Polyclonal Antibody
Anti-Matrix Metalloproteinase 15 Rabbit Polyclonal Antibody
Catalog # USBIM2430-03N
Supplier:  US Biological
Anti-Matrix Metalloproteinase 15 Rabbit Polyclonal Antibody
Catalog # USBIM2430-03N
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Matrix Metalloproteinase 15
  • Antigen symbol:
    MMP15
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2, 0,05% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recommended dilution: Immunochemistry: 1:50-1:100; Western Blot: 1:500-1:1000

Type: Primary
Antigen: Matrix Metalloproteinase 15
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human