Anti-MAP4K4 Rabbit Monoclonal Antibody [clone: 11C1046]
Catalog # USBIM2368-50
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Mitogen-activated protein kinase kinase kinase kinase 4
- Antigen symbol:MAP4K4
- Clonality:Monoclonal
- Clone:11C1046
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Monkey,Human,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:10mMsodium HEPES, pH 7,5, 150mM sodium chloride, 100ug/ml BSA, less than 0,02% sodium azide, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:1000
Type: Primary
Antigen: MAP4K4 (mitogen-activated protein kinase kinase kinase kinase 4)
Clonality: monoclonal
Clone: 11C1046
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey, Mouse