Order Entry
Export
Orders LinkContactUsLinkComponent
Anti-MAP2K6 Rabbit Polyclonal Antibody

Anti-MAP2K6 Rabbit Polyclonal Antibody

50 µg
Catalog # USBIM2363-22T
Supplier:   US Biological
Anti-MAP2K6 Rabbit Polyclonal Antibody
Catalog # USBIM2363-22T
Supplier:  US Biological
Anti-MAP2K6 Rabbit Polyclonal Antibody

50 µg

Price
390,00 €
Log in for availability
1 * 50 µG
Product Details & Documents

Recommended dilution: Immunofluorescence: 1:100-1:200; Western Blot: 1:500-1:1000; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: MAP2K6 (mitogen-activated protein kinase kinase 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat

Documents

Specifications
  • Size:
    50 µg
  • Western blot:
    Yes
  • Clonality:
    Polyclonal
  • Modification:
    phospho-Ser207
  • Purification:
    purified by immunoaffinity chromatography
  • Antibody type:
    Primary
  • Form:
    liquid
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Storage temperature:
    Cool
  • Host:
    Rabbit
  • Shipping temperature:
    Ice
  • Antigen name:
    Mitogen-activated protein kinase kinase 6
  • Cat. No.:
    USBIM2363-22T
  • Reactivity:
    Human|Rat
  • Conjugation:
    Unconjugated
  • Antigen symbol:
    MAP2K6
  • Storage buffer:
    PBS(without Mg2+ and Ca2+), pH 7,4, 150mMsodium chloride, 0,02% sodium azide, 50% glycerol.