Anti-MAP Kinase 15 Rabbit Polyclonal Antibody
Catalog # USBIM2362-65A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Mitogen-activated protein kinase 15
- Antigen symbol:MAPK15
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:1 to 245
- Storage buffer:0,1M Tris-glycine, pH 7, 0,01% thimerosal,10% glycerol.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:200; Immunochemistry: 1:100-1:200
Type: Primary
Antigen: MAP Kinase 15
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human