Anti-Lorazepam Sheep Polyclonal Antibody
Catalog # USBIL3428
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Lorazepam
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Sheep
- Form:liquid
- Storage buffer:20mMPhosphate, 150mM sodium chloride, 0,09% sodium Azide, pH 7,2.
- Storage temperature:Cool
- Concentration:~~1 mg/ml (UV abs)
- Shipping temperature:Ice
- Purification:purified
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: ELISA: 3.5 ?g/ml
Type: Primary
Antigen: Lorazepam
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: