Anti-Keratin 8, 18 Mouse Monoclonal Antibody [clone: NCL5D3]
USBIK0199-09Y
- Antibody type:Primary
- Antigen name:Keratin 8, 18
- Antigen symbol:KRT8, 18
- Clonality:Monoclonal
- Clone:NCL5D3
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a
- Western blot:Yes
- Format:ascites
- Form:liquid
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 ml
- Pk:1 mL
Recommended dilution: Immunochemistry: 1:5-1:10 is
Type: Primary
Antigen: Keratin 8, 18
Clonality: monoclonal
Clone: NCL5D3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: