Anti-IL34 Rabbit Polyclonal Antibody
100 µg
Catalog # USBII8443-93C
Supplier: US Biological
Jump to:
Product Details & Documents
Recommended dilution: Western Blot: 0.25-0.5 ?g/ml
Type: Primary
Antigen: IL34 (interleukin 34)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Documents
Specifications
- Size:100 µg
- Western blot:Yes
- Clonality:Polyclonal
- Purification:purified by immunoaffinity chromatography
- Antibody type:Primary
- Epitope:N-terminal
- Form:liquid
- Storage temperature:Cool
- Host:Rabbit
- Shipping temperature:Ice
- Antigen name:Interleukin 34
- Cat. No.:USBII8443-93C
- Reactivity:Human|Rat|Mouse
- Conjugation:Unconjugated
- Antigen symbol:IL34
- Storage buffer:PBS, 0,02% sodium azide.
- ELISA:Yes



