Anti-IFNL1 Rabbit Polyclonal Antibody (Biotin)
Catalog # USBII8443-78-BIOT
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Interferon lambda 1
- Antigen symbol:IFNL1
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: IFNL1 (interferon, lambda 1)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human