Anti-Interleukin 32 alpha Rat Monoclonal Antibody [clone: 11n107]
Catalog # USBII8442-44
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 32 alpha
- Antigen symbol:IL32A
- Clonality:Monoclonal
- Clone:11n107
- Conjugation:Unconjugated
- Host:Rat
- Isotype:IgG2a
- Reactivity:Human
- Form:lyophilized powder
- Amino acid number:3 to 131
- Storage buffer:PBS,trehalose.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A/G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 2?g/ml; Immunochemistry: 8-25 ?g/ml
Type: Primary
Antigen: Interleukin 32 alpha
Clonality: monoclonal
Clone: 11n107
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human