Anti-IL21R Rabbit Polyclonal Antibody
Catalog # USBII8442-23
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 21 Receptor
- Antigen symbol:IL21R
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,1% sodium azide before the addition of 40% glycerol.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: ELISA: 1:100000; Immunochemistry: 1:300
Type: Primary
Antigen: IL21R (interleukin 21 receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse