Order Entry
Export
ContactUsLinkComponent
Anti-Interleukin 20 Receptor alpha Rabbit Polyclonal Antibody
Anti-Interleukin 20 Receptor alpha Rabbit Polyclonal Antibody
Catalog # USBII8441-75
Supplier:  US Biological
Anti-Interleukin 20 Receptor alpha Rabbit Polyclonal Antibody
Catalog # USBII8441-75
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Interleukin 20 Receptor Alpha
  • Antigen symbol:
    IL20RA
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, 0,2% gelatin, 0,05% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: Interleukin 20 Receptor alpha
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse