Order Entry
Export
ContactUsLinkComponent
 
Anti-Interleukin 18 Rabbit Polyclonal Antibody
  USBII8439-95V
Anti-Interleukin 18 Rabbit Polyclonal Antibody
  USBII8439-95V

 

  • Antibody type:
    Primary
  • Antigen name:
    Interleukin 18
  • Antigen symbol:
    IL18
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, 0,1% BSA.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    1 ml
  • Pk:
    1 mL

 

 

Recommended dilution: Western Blot: 1:10

Type: Primary
Antigen: Interleukin 18
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse