Anti-Interleukin-17 Receptor A Goat Polyclonal Antibody
Catalog # USBII8439-36
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Interleukin-17 Receptor A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Rat
- Western blot:Yes
- Form:liquid
- Storage buffer:Tris saline, 0,02% sodium azide, pH 7,3, 0,5% BSA.
- Sequence:C-DQEKHGDDSKIN
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: 1:8000; Western Blot: 1-3?g/ml
Type: Primary
Antigen: Interleukin-17 Receptor A
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat