Anti-Interleukin 17 Receptor Mouse Monoclonal Antibody [clone: 7H43]
Catalog # USBII8439-35
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 17 Receptor
- Antigen symbol:IL17R
- Clonality:Monoclonal
- Clone:7H43
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:lyophilized powder
- Amino acid number:33 to 320
- Storage buffer:PBS, 5% trehalose.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:500 µg
- Pk:500 µG
Specifications
About this item
Recommended dilution: ELISA: 2-8?g/ml; Western Blot: 1-2?g/ml
Type: Primary
Antigen: Interleukin 17 Receptor
Clonality: monoclonal
Clone: 7H43
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human