Anti-Interleukin 17 Rabbit Polyclonal Antibody (Biotin)
Catalog # USBII8439-04P1
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 17
- Antigen symbol:IL17
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:lyophilized powder
- Storage buffer:PBS, pH 7,2. 500ul sterile 40-50% glycerol,0,1% BSA, PBS.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: ELISA: 0.25-1 ?g/ml; Western Blot: 0.1-0.2?g/ml
Type: Primary
Antigen: Interleukin 17
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse